Stampa…

2-Benzylacrylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Methylene-3-phenylpropionic Acid

B3409-25G B3409-5G
TCIAB3409-25GEA 48 EUR
TCIAB3409-25G TCIAB3409-5G
2-Benzylacrylic acid ≥98.0% (by GC, titration analysis)
2-Benzylacrylic acid
Formula: C₁₀H₁₀O₂
Peso molecolare: 162.19 g/mol
MDL Number: MFCD00854174
CAS Number: 5669-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72881849c4d9f924217644d15c7b9049cb9411a2&vid=72881849c4d9f924217644d15c7b9049cb9411a2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR