2,6-Bis(hydroxymethyl)naphthalene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Naphthalene-2,6-diyldimethanol
B3399-1G
B3399-5G
TCIAB3399-1GEA
93
EUR
TCIAB3399-1G
TCIAB3399-5G
2,6-Bis(hydroxymethyl)naphthalene ≥98.0% (by GC)
2,6-Bis(hydroxymethyl)naphthalene
Formula:
C₁₂H₁₂O₂ Peso molecolare: 188,23 g/mol |
MDL Number:
MFCD11870109 CAS Number: 5859-93-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 171.0 to 175.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...