Bis[2-(trimethylsilyloxy)ethyleEther ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Diethylene Glycol Bis(trimethylsilyl Ether)
B3378-25G
B3378-5G
TCIAB3378-25GEA
165
EUR
TCIAB3378-25G
TCIAB3378-5G
Bis[2-(trimethylsilyloxy)ethyleEther ≥98.0% (by GC)
Bis[2-(trimethylsilyloxy)ethyleEther
Formula:
C₁₀H₂₆O₃Si₂ Peso molecolare: 250,49 g/mol Punto di ebollizione: 87°C |
CAS Number:
16654-74-3 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...