Stampa…

Butyl-2-methylvalerate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Methylpentanoic Acid Butyl Ester, 2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate

B3359-25ML B3359-500ML
TCIAB3359-25MLEA 17 EUR
TCIAB3359-25ML TCIAB3359-500ML
Butyl-2-methylvalerate ≥98.0% (by GC)
Butyl-2-methylvalerate
Formula: C₁₀H₂₀O₂
Peso molecolare: 172,27 g/mol
CAS Number: 6297-41-2
EINECS: 228-569-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR