3-Bromo-5-methylpyridin-2-ol ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
B3350-1G
B3350-5G
TCIAB3350-1GEA
148
EUR
TCIAB3350-1G
TCIAB3350-5G
3-Bromo-5-methylpyridin-2-ol ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylpyridin-2-ol
Formula:
C6H6BrNO |
MDL Number:
MFCD03791312 CAS Number: 17282-02-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 158.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...