Stampa…

Bisdemethoxycurcumin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, 1,7-Bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione, Didemethoxycurcumin, Bisdemethoxycucurmin

B3347-25G B3347-5G
TCIAB3347-25GEA 245 EUR
TCIAB3347-25G TCIAB3347-5G
Bisdemethoxycurcumin ≥98.0% (by HPLC, titration analysis)
Bisdemethoxycurcumin
Formula: C₁₉H₁₆O₄
Peso molecolare: 308,33 g/mol
Punto di fusione: 224°C
MDL Number: MFCD03419284
CAS Number: 24939-16-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR