Stampa…

7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 7-(4-Bromobutoxy)-3,4-dihydroquinolin-2(1H)-one, 7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline

B3333-5G B3333-1G
TCIAB3333-5GEA 42 EUR
TCIAB3333-5G TCIAB3333-1G
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥98.0% (by HPLC, total nitrogen)
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone
Formula: C₁₃H₁₆BrNO₂
Peso molecolare: 298,18 g/mol
MDL Number: MFCD06658540
CAS Number: 129722-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe53784801e495c2dd8f923d4d2fea68d033806c&vid=fe53784801e495c2dd8f923d4d2fea68d033806c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR