4-Benzyloxy-1,3-butanediol ≥97.0% (by GC)
Fornitore: TCI
B3326-5G
B3326-1G
TCIAB3326-5GEA
842
EUR
TCIAB3326-5G
TCIAB3326-1G
4-Benzyloxy-1,3-butanediol ≥97.0% (by GC)
4-Benzyloxy-1,3-butanediol
Formula:
C₁₁H₁₆O₃ |
CAS Number:
71998-70-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...