Stampa…

1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane ≥94.0% (by GC)

Fornitore: TCI
Attenzione

B3319-25G B3319-5G
TCIAB3319-25GEA 100 EUR
TCIAB3319-25G TCIAB3319-5G
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane ≥94.0% (by GC)
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane
Formula: C₁₀H₂₁F₆NSi₂
Flash Pt: 78°C
MDL Number: MFCD00153070
CAS Number: 39482-87-6
EINECS: 254-470-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR