Stampa…

1-Boc-3-piperidone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-BOC-3-piperidone, 1-Boc-3-Piperidinone

B3309-5G B3309-25G
TCIAB3309-5GEA 58 EUR
TCIAB3309-5G TCIAB3309-25G
1-Boc-3-piperidone ≥98.0% (by GC)
1-Boc-3-piperidone
Formula: C₁₀H₁₇NO₃
Peso molecolare: 199,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Orange to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR