5-(4-Bromophenyl)-2-furaldehyde ≥94.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₁H₇BrO₂ |
MDL Number:
MFCD00239387 CAS Number: 20005-42-9 |
Risultati del test di specificazione
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (GC) | min. 94.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...