Stampa…

Butenafine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-(4-(tert-Butyl)benzyl)-N-methyl-1-(naphthalen-1-yl)methanamine hydrochloride, Butenafine HCl

B3293-1G B3293-5G
TCIAB3293-1GEA 35 EUR
TCIAB3293-1G TCIAB3293-5G
Butenafine hydrochloride ≥98.0% (by HPLC, titration analysis)
Butenafine hydrochloride
Formula: C₂₃H₂₇N
Peso molecolare: 353,93 g/mol
MDL Number: MFCD00917064
CAS Number: 101827-46-7
Merck Index: 14,01518

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 208.0 to 213.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR