Stampa…

1,2-Bis(phenylsulphinyl)ethanepalladium(II) diacetate ≥96.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: 1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst

B3292-1G B3292-200MG
TCIAB3292-1GEA 160 EUR
TCIAB3292-1G TCIAB3292-200MG
1,2-Bis(phenylsulphinyl)ethanepalladium(II) diacetate ≥96.0% (by titrimetric analysis)
1,2-Bis(phenylsulphinyl)ethanepalladium(II) diacetate
Formula: C₁₆H₁₈O₄PdS₂
Peso molecolare: 444.86 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD09842752
CAS Number: 858971-43-4

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Amber to Dark red powder to crystal
Purity (Chelometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR