O-Benzoyl-N-tert-butylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
B3238-1G
B3238-5G
TCIAB3238-1GEA
47
EUR
TCIAB3238-1G
TCIAB3238-5G
O-Benzoyl-N-tert-butylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
O-Benzoyl-N-tert-butylhydroxylamine hydrochloride
Formula:
C₁₁H₁₅NO₂ Punto di fusione: 187°C |
CAS Number:
66809-86-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...