2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Flecainide Impurity D
B3227-25G
B3227-5G
TCIAB3227-25GEA
78
EUR
TCIAB3227-25G
TCIAB3227-5G
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid
Formula:
C₁₁H₈F₆O₄ Peso molecolare: 318,17 g/mol |
MDL Number:
MFCD00221424 CAS Number: 35480-52-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...