4-Benzyl-L-aspartate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
L-Aspartic acid beta-benzyl ester, H-Asp(obzl)-OH, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, L-Aspartic Acid beta-Benzyl Ester
B3209-5G
B3209-25G
TCIAB3209-5GEA
17
EUR
TCIAB3209-5G
TCIAB3209-25G
4-Benzyl-L-aspartate ≥98.0% (by titrimetric analysis)
4-Benzyl-L-aspartate
Formula:
C₁₁H₁₃NO₄ Peso molecolare: 223,23 g/mol Punto di fusione: 221°C |
MDL Number:
MFCD00037208 CAS Number: 2177-63-1 EINECS: 218-541-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +24.0 ~ +30.0 deg (C=1, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...