Stampa…
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: (R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthalene, (R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
B3202-1G B3202-5G
TCIAB3202-1GEA 94 EUR
TCIAB3202-1G TCIAB3202-5G
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₂O₄
Peso molecolare: 374,429 g/mol
MDL Number: MFCD03788935
CAS Number: 173831-50-0
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C
Specific rotation [a]20/D +91.0 to +95.0 deg (C=1,CHCl3)