Stampa…

(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(tert-Butoxycarbonyl)-trans-4-fluoro-L-proline, (2S,4R)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, N-Boc-trans-4-fluoro-L-proline

B3177-1G B3177-200MG
TCIAB3177-1GEA 152 EUR
TCIAB3177-1G TCIAB3177-200MG
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥96.0% (by GC, titration analysis)
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Formula: C₁₀H₁₆FNO₄
Peso molecolare: 233,24 g/mol
MDL Number: MFCD06796085
CAS Number: 203866-14-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 96.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR