1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride ≥96.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
SIPr.HCl, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium chloride, 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride
B3157-1G
B3157-5G
B3157-500MG
TCIAB3157-1GEA
54
EUR
TCIAB3157-1G
TCIAB3157-5G
TCIAB3157-500MG
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride ≥96.0% (by HPLC, total nitrogen)
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride
Formula:
C₂₇H₃₉ClN₂ Peso molecolare: 427,07 g/mol |
MDL Number:
MFCD07369796 CAS Number: 258278-25-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...