4-Bromo-N,N-3-trimethylaniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Bromo-N,N-dimethyl-m-toluidine
B3133-25G
B3133-5G
TCIAB3133-25GEA
275
EUR
TCIAB3133-25G
TCIAB3133-5G
4-Bromo-N,N-3-trimethylaniline ≥98.0% (by GC, titration analysis)
4-Bromo-N,N-3-trimethylaniline
Formula:
C₉H₁₂BrN Peso molecolare: 214,11 g/mol Punto di ebollizione: 148°C |
MDL Number:
MFCD01851099 CAS Number: 50638-50-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...