Stampa…

1,4-Butanesultam ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 1,2-Thiazinane 1,1-Dioxide

B3117-1G B3117-5G
TCIAB3117-1GEA 125 EUR
TCIAB3117-1G TCIAB3117-5G
1,4-Butanesultam ≥98.0% (by GC)
1,4-Butanesultam
Formula: C₄H₉NO₂S
MDL Number: MFCD04038916
CAS Number: 37441-50-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR