Stampa…

N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥99.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Bz-DMT-dA, N-(9-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide

B3103-1G B3103-100MG
TCIAB3103-1GEA 197 EUR
TCIAB3103-1G TCIAB3103-100MG
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥99.0% (by HPLC)
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine
Formula: C₃₈H₃₅N₅O₆
Peso molecolare: 657,73 g/mol
Punto di fusione: 94°C
MDL Number: MFCD00010058
CAS Number: 64325-78-6
EINECS: 264-776-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -9.0 to -12.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR