Stampa…

N-Benzyl-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: N-Benzyl-p-toluenesulphonaMide, N-Tosylbenzylamine

B3082-25G B3082-5G
TCIAB3082-25GEA 216 EUR
TCIAB3082-25G TCIAB3082-5G
N-Benzyl-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N-Benzyl-p-toluenesulphonamide
Formula: C₁₄H₁₅NO₂S
Peso molecolare: 261,34 g/mol
MDL Number: MFCD00159328
CAS Number: 1576-37-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR