Stampa…

5-Bromo-2-iodopyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B3040-1G B3040-5G
TCIAB3040-1GEA 32 EUR
TCIAB3040-1G TCIAB3040-5G
5-Bromo-2-iodopyrimidine ≥98.0% (by GC)
5-Bromo-2-iodopyrimidine

This product is unavailable in the U.S.

Formula: C₄H₂BrIN₂
Peso molecolare: 284,88 g/mol
MDL Number: MFCD01318111
CAS Number: 183438-24-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR