N,N'-Bis(5-hydroxysalicylidene)ethylenediamine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Synonyms:
N,N'-Ethylenebis(5-hydroxysalicylideneimine), N,N-Bis(2,5-dihydroxybenzylidene)ethylenediamine, 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(benzene-1,4-diol)
Formula:
C₁₆H₁₆N₂O₄ Punto di fusione: 250°C |
MDL Number:
MFCD04039747 CAS Number: 124061-43-4 |
Risultati del test di specificazione
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...