Stampa…

1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: L-Aspartic Acid 1-Benzyl Ester, H-Asp-OBzl

B2994-1G B2994-5G
TCIAB2994-1GEA 18 EUR
TCIAB2994-1G TCIAB2994-5G
1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-L-aspartate
Formula: C₁₁H₁₃NO₄
Peso molecolare: 223,23 g/mol
MDL Number: MFCD00063186
CAS Number: 7362-93-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR