Stampa…

Bis(4-nitrophenyl)amine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-Dinitrodiphenylamine

B2960-1G B2960-5G
TCIAB2960-1GEA 84 EUR
TCIAB2960-1G TCIAB2960-5G
Bis(4-nitrophenyl)amine ≥98.0% (by HPLC)
Bis(4-nitrophenyl)amine
Formula: C₁₂H₉N₃O₄
Peso molecolare: 259,22 g/mol
MDL Number: MFCD00437044
CAS Number: 1821-27-8
EINECS: 217-343-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 215.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR