Stampa…

3,5-Bis(trifluoromethyl)benzylamine ≥95.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

B2949-1G B2949-5G
TCIAB2949-1GEA 32 EUR
TCIAB2949-1G TCIAB2949-5G
3,5-Bis(trifluoromethyl)benzylamine ≥95.0% (by GC)
3,5-Bis(trifluoromethyl)benzylamine
Formula: C₉H₇F₆N
Peso molecolare: 243.15 g/mol
MDL Number: MFCD00009909
CAS Number: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8 II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR