Stampa…

2-(3-Benzoylphenyl)propionitrile ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B2939-25G B2939-5G
TCIAB2939-25GEA 183 EUR
TCIAB2939-25G TCIAB2939-5G
2-(3-Benzoylphenyl)propionitrile ≥98.0% (by GC)
2-(3-Benzoylphenyl)propionitrile
Formula: C₁₆H₁₃NO
MDL Number: MFCD00044435
CAS Number: 42872-30-0
EINECS: 255-982-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR