Stampa…

2-Methylpropane-2-sulphinamide ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: tert-Butylsulphinamide

B2926-1G B2926-5G
TCIAB2926-1GEA 28 EUR
TCIAB2926-1G TCIAB2926-5G
2-Methylpropane-2-sulphinamide ≥97.0% (by GC)
2-Methylpropane-2-sulphinamide
Formula: C₄H₁₁NOS
Peso molecolare: 121,20 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01863616
CAS Number: 146374-27-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR