tert-Butyldiphenylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
tert-Butyldiphenylsilyl Triflate, Trifluoromethanesulphonic acid tert-Butyldiphenylsilyl Ester
B2898-1G
B2898-5G
TCIAB2898-1GEA
85
EUR
TCIAB2898-1G
TCIAB2898-5G
tert-Butyldiphenylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
tert-Butyldiphenylsilyl trifluoromethanesulphonate
Formula:
C₁₇H₁₉F₃O₃SSi Peso molecolare: 388,48 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01941212 CAS Number: 92886-86-7 UN: 1760 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Brown clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...