Stampa…

Bis(3-amino-4-hydroxyphenyl)sulphone ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,3'-Diamino-4,4'-dihydroxydiphenyl sulphone, 3,3'-Diamino-4,4'-dihydroxydiphenyl Sulphone

B2859-5G B2859-25G
TCIAB2859-5GEA 108 EUR
TCIAB2859-5G TCIAB2859-25G
Bis(3-amino-4-hydroxyphenyl)sulphone ≥98.0% (by HPLC)
Bis(3-amino-4-hydroxyphenyl)sulphone
Formula: C₁₂H₁₂N₂O₄S
Peso molecolare: 280,30 g/mol
Punto di fusione: 231°C
MDL Number: MFCD00025181
CAS Number: 7545-50-8
EINECS: 231-428-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR