Stampa…

tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate ≥95.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: O-[Bis(4-methoxyphenyl)phosphinyl]-N-Boc-hydroxylamine, O-[Bis(4-methoxyphenyl)phosphinyl]-N-(tert-butoxycarbonyl)hydroxylamine, [Bis(4-methoxyphenyl)phosphinyloxy]carbamic Acid tert-Butyl Ester

B2857-1G B2857-5G
TCIAB2857-1GEA 174 EUR
TCIAB2857-1G TCIAB2857-5G
tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate ≥95.0% (by HPLC, total nitrogen)
tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate
Formula: C₁₉H₂₄NO₆P
Peso molecolare: 393,38 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 619333-95-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR