Stampa…

4-Bromo-4'-(diphenylamino)biphenyl ≥93.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4'-Bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine, (4'-Bromobiphenyl-4-yl)diphenylamine

B2844-1G B2844-250MG
TCIAB2844-1GEA 384 EUR
TCIAB2844-1G TCIAB2844-250MG
4-Bromo-4'-(diphenylamino)biphenyl ≥93.0% (by GC)
4-Bromo-4'-(diphenylamino)biphenyl
Formula: C₂₄H₁₈BrN
Peso molecolare: 400,32 g/mol
MDL Number: MFCD09038499
CAS Number: 202831-65-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 93.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR