Stampa…

2-Butyl-2-adamantanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Butyl-2-hydroxyadamantane

B2839-5G B2839-25G
TCIAB2839-5GEA 88 EUR
TCIAB2839-5G TCIAB2839-25G
2-Butyl-2-adamantanol ≥98.0% (by GC)
2-Butyl-2-adamantanol
Formula: C₁₄H₂₄O
Peso molecolare: 208,35 g/mol
CAS Number: 14451-86-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR