Stampa…

Betulinic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

B2836-100MG B2836-1G
TCIAB2836-100MGEA 48 EUR
TCIAB2836-100MG TCIAB2836-1G
Betulinic acid ≥97.0% (by GC, titration analysis)
Betulinic acid
Formula: C₃₀H₄₈O₃
Peso molecolare: 456,71 g/mol
MDL Number: MFCD00009619
CAS Number: 472-15-1
EINECS: 207-448-8
Merck Index: 14,01190

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 % (after drying)
Specific rotation [a]20/D +6.0 to +10.0 deg (C=0.9, pyridine)
Drying loss max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR