Benzyl-6-bromohexyl ether ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(((6-Bromohexyl)oxy)methyl)benzene, Benzyl 6-Bromohexyl Ether, Benzyloxyhexyl Bromide, (6-Bromohexyloxymethyl)benzene
B2820-25G
B2820-5G
TCIAB2820-25GEA
387
EUR
TCIAB2820-25G
TCIAB2820-5G
Benzyl-6-bromohexyl ether ≥97.0% (by GC)
Benzyl-6-bromohexyl ether
Formula:
C₁₃H₁₉BrO Peso molecolare: 271,20 g/mol |
MDL Number:
MFCD02258455 CAS Number: 54247-27-7 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...