Stampa…

6-(Benzyloxy)-1H-indole ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 6-Benzyloxyindole

B2809-25G B2809-5G B2809-1G
TCIAB2809-25GEA 530 EUR
TCIAB2809-25G TCIAB2809-5G TCIAB2809-1G
6-(Benzyloxy)-1H-indole ≥98.0% (by GC)
6-(Benzyloxy)-1H-indole
Formula: C15H13NO
MDL Number: MFCD00053554
CAS Number: 15903-94-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR