Stampa…

9-Benzylcarbazole-3-carboxaldehyde ≥97.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-Benzyl-9H-carbazole-3-carbaldehyde, 9-Benzyl-3-formylcarbazole

B2804-1G B2804-5G
TCIAB2804-1GEA 91 EUR
TCIAB2804-1G TCIAB2804-5G
9-Benzylcarbazole-3-carboxaldehyde ≥97.0% (by total nitrogen basis)
9-Benzylcarbazole-3-carboxaldehyde
Formula: C₂₀H₁₅NO
Peso molecolare: 285,35 g/mol
MDL Number: MFCD00403522
CAS Number: 54117-37-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 131.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR