6-Bromoindole-3-carboxaldehyde ≥98.0% (by GC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromo-3-formylindole, 6-Bromo-1H-indole-3-carbaldehyde
B2778-1G
B2778-5G
TCIAB2778-1GEA
41
EUR
TCIAB2778-1G
TCIAB2778-5G
6-Bromoindole-3-carboxaldehyde ≥98.0% (by GC, total nitrogen)
6-Bromoindole-3-carboxaldehyde
Formula:
C₉H₆BrNO Peso molecolare: 224,06 g/mol |
MDL Number:
MFCD00792689 CAS Number: 17826-04-9 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 201.0 to 206.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...