Stampa…

4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2755-25G B2755-5G
TCIAB2755-25GEA 101 EUR
TCIAB2755-25G TCIAB2755-5G
4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine ≥98.0% (by GC)
4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine
Formula: C₇H₆F₄N₂O₂S
Peso molecolare: 258,19 g/mol
CAS Number: 100478-25-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR