(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
B2709-1G
B2709-5G
B2709-25G
TCIAB2709-1GEA
31
EUR
TCIAB2709-1G
TCIAB2709-5G
TCIAB2709-25G
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0% (by HPLC)
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
Formula:
C₃₉H₃₂OP₂ Peso molecolare: 578,62 g/mol |
MDL Number:
MFCD00233866 CAS Number: 161265-03-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 227.0 to 231.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...