Stampa…

Bis(4-iodophenyl)amine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B2685-1G B2685-5G
TCIAB2685-1GEA 84 EUR
TCIAB2685-1G TCIAB2685-5G
Bis(4-iodophenyl)amine ≥98.0% (by GC)
Bis(4-iodophenyl)amine
Formula: C₁₂H₉I₂N
MDL Number: MFCD00454093
CAS Number: 20255-70-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR