Stampa…

Bis(4-bromophenyl)amine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-Dibromodiphenylamine

B2684-1G B2684-5G
TCIAB2684-1GEA 16 EUR
TCIAB2684-1G TCIAB2684-5G
Bis(4-bromophenyl)amine ≥98.0% (by GC)
Bis(4-bromophenyl)amine
Formula: C₁₂H₉Br₂N
Peso molecolare: 327,02 g/mol
MDL Number: MFCD00225488
CAS Number: 16292-17-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR