Stampa…

4-(trans-4-Butylcyclohexyl)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-(Trans-4-butylcyclohexyl)phenol, 1-(trans-4-Butylcyclohexyl)-4-hydroxybenzene

B2644-25G B2644-100G
TCIAB2644-25GEA 64 EUR
TCIAB2644-25G TCIAB2644-100G
4-(trans-4-Butylcyclohexyl)phenol ≥98.0% (by GC)
4-(trans-4-Butylcyclohexyl)phenol
Formula: C₁₆H₂₄O
Peso molecolare: 232,37 g/mol
MDL Number: MFCD00673752
CAS Number: 88581-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c6b194baec100d2d20d9b6f52a8cfe98610f340&vid=8c6b194baec100d2d20d9b6f52a8cfe98610f340&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR