2,2'-Bis(diphenylphosphino)biphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
BIPHEP, 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl
B2630-1G
B2630-100MG
TCIAB2630-1GEA
146
EUR
TCIAB2630-1G
TCIAB2630-100MG
2,2'-Bis(diphenylphosphino)biphenyl ≥98.0% (by GC)
2,2'-Bis(diphenylphosphino)biphenyl
Formula:
C₃₆H₂₈P₂ Peso molecolare: 522,57 g/mol |
MDL Number:
MFCD03094574 CAS Number: 84783-64-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...