Bis[3-(trimethoxysilyl)propyl]amine ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₂H₃₁NO₆Si₂ |
MDL Number:
MFCD00191835 CAS Number: 82985-35-1 EINECS: 280-084-5 UN: 3082 ADR: 9,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...