1,4-Bis(trimethylsilyl)benzene ≥97.0% (by GC)
Fornitore: TCI
Formula:
C₁₂H₂₂Si₂ Punto di ebollizione: 194°C |
MDL Number:
MFCD00015592 CAS Number: 13183-70-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...