Stampa…

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98.0% (by GC)

Fornitore: TCI

B2436-100MG B2436-500MG
TCIAB2436-100MGEA 102 EUR
TCIAB2436-100MG TCIAB2436-500MG
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98.0% (by GC)
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene
Formula: C₁₈H₂₈P₂
MDL Number: MFCD00142336
CAS Number: 147253-67-6
Merck Index: 14,03466

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -470 to -520 deg (C=1, hexane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR