Stampa…

Biotin hydrazide ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide

B2431-25MG B2431-100MG
TCIAB2431-25MGEA 26 EUR
TCIAB2431-25MG TCIAB2431-100MG
Biotin hydrazide ≥98.0% (by HPLC)
Biotin hydrazide
Formula: C₁₀H₁₈N₄O₂S
Peso molecolare: 258,34 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00078532
CAS Number: 66640-86-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Protein labeling test (qualitative methoed) to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR